Order Entry
Export
Orders LinkContactUsLinkComponent
Ammonium acetate in aqueous solution

Ammonium acetate in aqueous solution

Glass bottle, 100 mL
Catalog # AMBI9070G
Supplier:   Ambion
Ammonium acetate in aqueous solution
Catalog # AMBI9070G
Supplier:  Ambion
CAS Number:  631-61-8
Ammonium acetate in aqueous solution
Ammonium acetate in aqueous solution

Glass bottle, 100 mL

CAS Number:  631-61-8
Price
Inquire For Price
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  Acetic acid ammonium salt
  • CAS Number:631-61-8
  • Formula:H₃CCOONH₄
  • MDL Number:MFCD00013066
  • Molecular Weight:77.08 g/mol
  • Storage Temperature:Ambient
  • EINECS:211-162-9
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications