Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
NorthernMax Low Stringency Wash Buffer 1 * 1 l
NorthernMax Low Stringency Wash Buffer 1 * 1 l
Print
NorthernMax Low Stringency Wash Buffer 1 * 1 l
Catalog # AMBI8673
Supplier:
Ambion
NorthernMax Low Stringency Wash Buffer 1 * 1 l
Catalog # AMBI8673
Supplier:
Ambion
Expand image
Return to Top
NorthernMax Low Stringency Wash Buffer 1 * 1 l
Add to cart
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 1 L
-
+
Inquire For Price
Return Policy
Product Details & Documents