Orders LinkContactUsLinkComponent
Accessories for epMotion®
Accessories for epMotion®
Thermal module for heating/cooling microplates and thermal racks from 0 to +110 °C (only for epMotion® 5075 automated pipetting systems)
Catalog # 613-9243
Supplier:   EPPENDORF
Accessories for epMotion®
Catalog # 613-9243
Supplier:  EPPENDORF
Accessories for epMotion®
Accessories for epMotion®
Thermal module for heating/cooling microplates and thermal racks from 0 to +110 °C (only for epMotion® 5075 automated pipetting systems)
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Thermal module for heating/cooling microplates and thermal racks from 0 to +110 °C (only for epMotion® 5075 automated pipetting systems)
  • Seq:
    33