Order Entry
Export
Orders LinkContactUsLinkComponent
Pectin, powder

Pectin, powder

1 kg
Catalog # ICNA0210258791
Supplier:   MP Biomedicals
Pectin, powder
Catalog # ICNA0210258791
Supplier:  MP Biomedicals
CAS Number:  9000-69-5
Pectin, powder

1 kg

CAS Number:  9000-69-5
Price
661,00 €
Log in for availability
1 * 1 kg
Product Details & Documents

Soluble in water (1 g/20 ml - viscous solution acid to litmus); insoluble in organic solvents.

Pectin is a heterosaccharide, component of terrestrial plant cell walls.

It is used as a substrate to identify, differentiate and characterised pectinase(s). It is also used to study its degradation by pectinolytic bacteria.

  • CAS Number:9000-69-5
  • MDL Number:MFCD00081838

Documents

Specifications

Specifications

  • Pk:
    1 kg

Specifications