Order Entry
Export
Orders LinkContactUsLinkComponent
L-Methionine

L-Methionine

Glass bottle for solids, 500 g
Catalog # ICNA02102291.5
Supplier:   MP Biomedicals
L-Methionine
Catalog # ICNA02102291.5
Supplier:  MP Biomedicals
CAS Number:  63-68-3
L-Methionine

Glass bottle for solids, 500 g

CAS Number:  63-68-3
Price
308,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
  • CAS Number:63-68-3
  • Formula:C₅H₁₁NO₂S
  • MDL Number:MFCD00063097
  • Molecular Weight:149.21 g/mol
  • Merck Index:13,06004
  • Boiling Pt:278 °C (1013 hPa)
  • Melting Pt:136 °C
  • Density:1.373 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-562-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications