Order Entry
Export
Orders LinkContactUsLinkComponent
AccuGENE® 1M Tris HCl buffer

AccuGENE® 1M Tris HCl buffer

AccuGENE™ 1 M Tris HCl buffer, 7,4, 1 L
Catalog # 733-1653
Supplier:   LONZA
AccuGENE® 1M Tris HCl buffer
Catalog # 733-1653
Supplier:  LONZA
CAS Number:  
AccuGENE® 1M Tris HCl buffer

AccuGENE™ 1 M Tris HCl buffer, 7,4, 1 L

CAS Number:  
Price
86,80 €
Log in for availability
1 * 1 L
Product Details & Documents

AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.

  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
  • Efficient - Ready-made solutions eliminate experiment preparation time
  • Flexible - Customized solutions are available to meet individual needs

Tris-HCl is a solution containing tris base mixed with hydrocholoric acid. Tris has a pKa of 8,07 at STP and is used in bufferings solutions to prevent large changes in pH level. A staple of electrophoretic and other biological applications, Tris has a useful buffer range of 7 to 9, which coincides with the physiological pH typical of most living organisms. Tris is the 'T' in TAE and TBE Buffers.

Each reagent is prepared with 18 megOhm water, filtered using a 0,2 micron filter, and filled into sterile bottles.

Store all reagents at 18 to 26°C

Documents

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    AccuGENE™ 1 M Tris HCl buffer
  • pH value:
    7,4

Specifications