Polarising microscope, PM-1805
Widefield cross reticle eyepiece
Catalog # 630-1169
Supplier: MOTIC
Jump to:
Product Details & Documents
Polarising microscope for the observation of fibres and plastics.
- Circular stage graduated from 0 to 360°
- Swing-in/out analyser and polariser
Delivery information: Supplied with a vinyl dust cover.
Documents
EDUCATION_LINE_18_28_PM_CA_1505_EN
Product Literature
Specifications
- Magnification:10×/18 mm
- Description:Widefield cross reticle eyepiece
- Seq:102
Accessories
Product Information
- Catalog #MagnificationAvailabilityPrice
- Micrometer eyepiece, 100 divisions in 10 mmMicrometer eyepiece, 100 divisions in 10 mm632-894610×/18Log in for availability1 * 1 items - 65,00 €
Product Information
- Catalog #MagnificationAvailabilityPrice
- Widefield cross reticle eyepiece