Orders LinkContactUsLinkComponent
Electroporation plates
Electroporation plates
Electroporation plates, 4 mm gap, 96, 250 µl
Catalog # 732-0477
Supplier:   BTX
Electroporation plates
Catalog # 732-0477
Supplier:  BTX
Electroporation plates
Electroporation plates
Electroporation plates, 4 mm gap, 96, 250 µl
Price
320,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Sterile multi-well electroporation plates for transfection/transformation of bacteria, yeast, insect, plant and mammalian cells.
Specifications
  • No. of wells:
    96
  • Volume:
    250 µl
  • Description:
    Electroporation plates, 4 mm gap