Orders LinkContactUsLinkComponent
Electroporation plates
Electroporation plates
Electroporation plates, 2 mm gap, 25, 125 µl
Catalog # 732-0481
Supplier:   BTX
Electroporation plates
Catalog # 732-0481
Supplier:  BTX
Electroporation plates
Electroporation plates
Electroporation plates, 2 mm gap, 25, 125 µl
Price
605,00 €
Log in for availability
1 * 6 items
Product Details & Documents
Sterile multi-well electroporation plates for transfection/transformation of bacteria, yeast, insect, plant and mammalian cells.
Specifications
  • No. of wells:
    25
  • Volume:
    125 µl
  • Description:
    Electroporation plates, 2 mm gap