pH buffers, DuraCal
9,21, ±0,02, Blue, Plastic bottle, 250 mL
Supplier: HAMILTON BONADUZ
Jump to:
Product Details & Documents
For full information concerning health and safety data please contact Avantor or Hamilton directly.
- Label includes expiration date, batch number and actual value
- Traceable to NIST and PTB, certificates available to download
- Stable solutions
- Certified pH value from a DAkkS laboratory accredited for pH measurement
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:250 mL
- Colour:Blue
- pH accuracy:±0,02
- pH value:9,21



