Screwthread tubes, attachable, DURAN®
GL 14, 12 mm, 1,5 mm
SCOT286251603
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.
: These tubes are open at the bottom.
- Thread:GL 14
- Tube ext.Ø:12 mm
- Wall thickness:1,5 mm
- Seq:01



