Stopcocks, single-way, DURAN®
6 mm, 21,5 NS
Catalog # SCOT286407409
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
- Replaceable NS keys
- Screw thread retaining device
Straight bore.
Specifications
- Hole Ø:6 mm
- Seq:02
- Spigot:21,5 NS