Stopcocks, single-way, DURAN®
4 mm, 18,8 NS
Catalog # SCOT286406807
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
- Replaceable NS keys
- Screw thread retaining device
Straight bore.
Specifications
- Hole Ø:4 mm
- Seq:13
- Spigot:18,8 NS