Orders LinkContactUsLinkComponent
Lids for Nunc™ microplates
Lid, PS, with condensation rings and evaporation barrier, for use with LowBot™ 384- and 1536-well plates, Clear, 4,6 mm, Sterile
Catalog # NUNC253624
   Low Manufacturing Impact
Lids for Nunc™ microplates
Catalog # NUNC253624
   Low Manufacturing Impact
Lids for Nunc™ microplates
Lids for Nunc™ microplates
Lid, PS, with condensation rings and evaporation barrier, for use with LowBot™ 384- and 1536-well plates, Clear, 4,6 mm, Sterile
Price
582,00 €
Log in for availability
1 * 100 items
Specifications
  • Sterility:
    Sterile
  • Colour:
    Clear
  • Description:
    Lid, PS, with condensation rings and evaporation barrier, for use with LowBot™ 384- and 1536-well plates
  • Height:
    4,6 mm
  • Seq:
    03
  • Cat. No.:
    NUNC253624
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact