Microplates, opaque OptiPlates™
OptiiPlate™-96 microplates, 96, White, Case of 50
Catalog # PERK6005290
Supplier: PerkinElmer
Jump to:
Product Details & Documents
These plates are used for general isotopic, fluorescence, LANCE™, [FP]², AlphaScreen™ or luminescence assays.
- Made of polystyrene
Specifications
- No. of wells:96
- Packed:Case of 50
- Colour:White
- Description:OptiiPlate™-96 microplates
- Seq:02