Order Entry
Export
Orders LinkContactUsLinkComponent
4'-Ethoxyacetanilide ≥97%

4'-Ethoxyacetanilide ≥97%

Plastic bottle for solids, 500 g
Catalog # A11200.36
Health
Irritant
4'-Ethoxyacetanilide ≥97%
Catalog # A11200.36
CAS Number:  62-44-2
4'-Ethoxyacetanilide ≥97%
4'-Ethoxyacetanilide ≥97%

Plastic bottle for solids, 500 g

CAS Number:  62-44-2
Price
88,10 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 97% CAS No.: 62-44-2 Molecular Formula: C10H13NO2 Formula Weight: 179.22 Melting Point: 134-136° Beilstein Reference: 1869238 Merck Reference: 14,7204 MDL No.: MFCD00009094

Synonyms:  Acetphenetidin, Acetophenetidin, Phenacetin, N-(4-Ethoxyphenyl)acetamide, 4-Acetophenetidide
  • CAS Number:62-44-2
  • Formula:C₁₀H₁₃NO₂
  • MDL Number:MFCD00009094
  • Molecular Weight:179.22 g/mol
  • Merck Index:14,7204
  • Melting Pt:134…136 °C
  • Storage Temperature:Ambient
  • EINECS:200-533-0
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications