Orders LinkContactUsLinkComponent
DPD chlorine testing systems, SwifTest™
DPD chlorine testing systems, SwifTest™
Refill vial for SwifTest™ dispenser DPD total chlorine reagent, 0,02 - 2,00 mg/l, DPD, SW, 250 Tests
Catalog # HACH2105660
Supplier:   Hach
DPD chlorine testing systems, SwifTest™
Catalog # HACH2105660
Supplier:  Hach
CAS Number:  
DPD chlorine testing systems, SwifTest™
DPD chlorine testing systems, SwifTest™
Refill vial for SwifTest™ dispenser DPD total chlorine reagent, 0,02 - 2,00 mg/l, DPD, SW, 250 Tests
CAS Number:  
Price
83,30 €
Log in for availability
1 * 250 Tests
Product Details & Documents
  • Fast, easy reagent addition - ideal for high volume testing
  • Just squeeze to add powder, no need to open reagent containers
Delivery information: Dispenser is supplied with 1 refill vial with enough reagent for approximately 250 tests.
Specifications
Specifications
  • Chemical method:
    DPD
  • Pk:
    250 Tests
  • Description:
    Refill vial for SwifTest™ dispenser DPD total chlorine reagent
  • Type:
    SW
  • Range:
    0,02 - 2,00 mg/l
Specifications