Order Entry
Export
Orders LinkContactUsLinkComponent
Sample vials, trident vials, squat or tall

Sample vials, trident vials, squat or tall

Squat, 10,50 ml, 23×46 mm, Yes
Catalog # 212-7066
Supplier:   MURRAY S
Sample vials, trident vials, squat or tall
Catalog # 212-7066
Supplier:  MURRAY S
Sample vials, trident vials, squat or tall
Sample vials, trident vials, squat or tall

Squat, 10,50 ml, 23×46 mm, Yes

Price
155,00 €
Log in for availability
1 * 190 items
Product Details & Documents

Vials made from neutral glass with PE plug stoppers. Suitable for a wide variety of uses in the laboratory, production, more.

  • Robust and economical, will withstand moderate temperature change
  • Wide opening for easy filling and emptying
  • Plug stopper for secure closure

Packaging: For production related reasons, the number of vials per box may vary.

Specifications
  • Cap included:
    Yes
  • Ø×H:
    23×46 mm
  • Capacity:
    10,50 ml
  • Description:
    Squat