Flat flange lids, centre neck
laboratory flange DN 100, 1 Neck, NS 29/32 (centre), 95 mm
Catalog # LENZ06100204
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Type:1 Neck
- With:laboratory flange DN 100
- Neck:NS 29/32 (centre)
- Total height:95 mm