Order Entry
Export
Orders LinkContactUsLinkComponent
 

Methanol ≥98.5%, TECHNICAL

Metal drum, 189 L
  20903.550
 
 
 
   Safer Human and Environmental Health
Methanol ≥98.5%, TECHNICAL
  20903.550
   Safer Human and Environmental Health
 :  67-56-1
Methanol ≥98.5%, TECHNICAL
Methanol ≥98.5%, TECHNICAL

Metal drum, 189 L

 :  67-56-1
 
912,00 €
   
1 * 189 L
 
 :  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  •  :67-56-1
  •  :CH₃OH
  •  :MFCD00004595
  •  :32.04 g/mol
  •  :13,05984
  •  :64.6 °C (1013 hPa)
  •  :–98 °C
  •  :0.7918 g/cm³ (20 °C)
  •  :11 °C (closed cup)
  •  :Ambient
  •  :200-659-6
  •  :1230
  •  :3 (6.1),II
  •  :01-2119433307-44
 

 

  • Assay:
    Min. 98.5 %
  • Evaporation residue:
    Max. 0.015 %

 

  • Pack type:
    Metal drum
  • Pk:
    189 L
  • Cat. No.:
    20903.550
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health