Orders LinkContactUsLinkComponent
β-glucuronidase substrates
β-glucuronidase substrates
Rose-β-D-GlcU,CHA, 25 mg
Catalog # 10019.
Supplier:   Biotium
β-glucuronidase substrates
Catalog # 10019.
Supplier:  Biotium
CAS Number:  
β-glucuronidase substrates
β-glucuronidase substrates
Rose-β-D-GlcU,CHA, 25 mg
CAS Number:  
Price
261,00 €
Log in for availability
1 * 25 mg
Product Details & Documents
β-glucuronidase substrates are easy to detect stains against the background of plant cells.
White solid soluble in water or DMSO.
Caution: Store at −20 °C and protect from light.
Specifications
Specifications
  • Pk:
    25 mg
  • Description:
    Rose-β-D-GlcU,CHA
Specifications