Orders LinkContactUsLinkComponent
β-glucuronidase substrates
Methylumbelliferyl-β-D-glucuronic acid dihydrate, 100 mg
  10020.
 :   Biotium
β-glucuronidase substrates
  10020.
 :  Biotium
 :  
β-glucuronidase substrates
β-glucuronidase substrates
Methylumbelliferyl-β-D-glucuronic acid dihydrate, 100 mg
 :  
180,00 €
Log in for availability
1 * 100 mg
β-glucuronidase substrates are easy to detect stains against the background of plant cells.
White solid soluble in water or DMSO.
 : Store at −20 °C and protect from light.
Specifications
Specifications
  • Pk:
    100 mg
  • Description:
    Methylumbelliferyl-β-D-glucuronic acid dihydrate
Specifications