Order Entry
Export
Orders LinkContactUsLinkComponent
Beta-galactosidase substrates

Beta-galactosidase substrates

β-D-galactopyranoside, Green, 25 mg
Catalog # 10015.
Supplier:   Biotium
Beta-galactosidase substrates
Catalog # 10015.
Supplier:  Biotium
CAS Number:  
Beta-galactosidase substrates
Beta-galactosidase substrates

β-D-galactopyranoside, Green, 25 mg

CAS Number:  
Price
267,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Chromogenic and fluorogenic beta-galactosidase substrates.

White solid form soluble in water and DMSO.

Ordering information: Includes 5 mg of X-gal, red-beta-D-gal, rose-beta-D-gal, purple-beta-D-gal and green-beta-D-gal.

Caution: Store at −20 °C and protect from light.

Specifications

Specifications

  • Pk:
    25 mg
  • Colour:
    Green
  • Description:
    β-D-galactopyranoside
  • Cat. No.:
    10015.
  • Storage conditions:
    Protect from light
  • Storage temperature:
    −20 °C

Specifications