Orders LinkContactUsLinkComponent
Connectors
Connectors
Fittings, external thread for pipes and valves, 1/2"
Catalog # SEMA4366
Supplier:   SEMADENI
Connectors
Catalog # SEMA4366
Supplier:  SEMADENI
Connectors
Connectors
Fittings, external thread for pipes and valves, 1/2"
Price
26,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Connectors for easy installation on any plastic containers by means of counter screw.
  • Male thread for fittings or hose connection
  • Good chemical resistance to acids and solvents
Specifications
  • Thread:
    1/2"
  • Description:
    Fittings, external thread for pipes and valves
  • Seq:
    01