Orders LinkContactUsLinkComponent
Screw caps for screw thread tubes
Screw caps for screw thread tubes
GL 14
Catalog # LENZ01320014
Supplier:   Lenz Laborglas
Screw caps for screw thread tubes
Catalog # LENZ01320014
Supplier:  Lenz Laborglas
Screw caps for screw thread tubes
Screw caps for screw thread tubes
GL 14
Price
3,00 €
Log in for availability
1 * 1 items
Product Details & Documents
PBT,red, with PTFE coated silicone sealing (VMQ).
  • For DIN GL threads
  • Heat resistant up to 180 °C
  • Autoclavable, sterilisable with hot air
Specifications
  • Thread:
    GL 14
  • Seq:
    01