Order Entry
Export
Orders LinkContactUsLinkComponent
Test tube baskets

Test tube baskets

225×225×230 mm
Catalog # 215-0550
Supplier:   SciLabware
Test tube baskets
Catalog # 215-0550
Supplier:  SciLabware
Test tube baskets
Test tube baskets

225×225×230 mm

Price
30,90 €
Log in for availability
1 * 1 items
Product Details & Documents

PP.

  • Very good chemical resistance
  • Autoclavable at 135 °C
  • Highly break-resistant
  • Stackable space saving design

These tests tubes baskets are suitable for rinsing, sterilising, transporting and storing glassware.

Colour: White

Specifications
  • W×D×H:
    225×225×230 mm