Volumetric flasks, class B, Azlon®
250 ml, 19/26, 0,30 ± ml
214-0118
PP, translucent, with PE stopper.
- Class B tolerances with a clear easy to read calibration line
- Feature a mixing chamber at the top of each flask's neck
- Exposure to temperatures above 60 °C may affect accuracy
- Cleaning with mild alkaline detergents recommended up to 60 °C
: ISO 1042
- NS:19/26
- Capacity:250 ml
- Tolerance:0,30 ± ml



