Order Entry
Export
Orders LinkContactUsLinkComponent
 

Volumetric flasks, class B, Azlon®

25 ml, 10/19, 0,08 ± ml
  214-0115
 :   SciLabware
Volumetric flasks, class B, Azlon®
  214-0115
Volumetric flasks, class B, Azlon®
Volumetric flasks, class B, Azlon®

25 ml, 10/19, 0,08 ± ml

 
22,50 €
   
1 * 1 items
 

PP, translucent, with PE stopper.

  • Class B tolerances with a clear easy to read calibration line
  • Feature a mixing chamber at the top of each flask's neck
  • Exposure to temperatures above 60 °C may affect accuracy
  • Cleaning with mild alkaline detergents recommended up to 60 °C

 : ISO 1042

 
  • NS:
    10/19
  • Capacity:
    25 ml
  • Tolerance:
    0,08 ± ml