Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Bottle, narrow mouth LDPE with PP cap 250 mL 1 * 10 items
Catalog # 215-0513
Supplier:
SciLabware
Expand image
Bottle, narrow mouth LDPE with PP cap 250 mL 1 * 10 items
Catalog # 215-0513
Supplier:
SciLabware