VWR®, Combined pH Electrodes, pHASE
Sealed, double junction (Pellon), 0 - 14, Glass, Gel, 12×150 mm, BNC, Aqueous media, beverage, Tris buffer
Catalog # 662-1799
Supplier: VWR Collection
Jump to:
Product Details & Documents
Combined electrodes providing fast, accurate and stable pH measurements, ideal for control quality and research and development use.
- Fast, accurate and stable pH measurements, fast thermal equilibration, high accuracy even in samples with large temperature range
- Available with sealed design (economical, easy to use, with minimum maintenance) or Rapid-Renew design (refillable electrodes used with dirty or other samples that cause junction fouling)
- Available with epoxy body (with good chemical resistance, maximum ruggedness and removable bulb guard) or glass body (maximum chemical resistance with fully exposed ruggedised bulbs)
- With 1 m fixed cable
pH range: 0 to 14
Accuracy: to 0,04 pH between 0 to 100 °C in pH 7 buffer
Stability: 0,005 pH or less drift per hour
Temperature range: 0 to +100 °C
Temperature range: 0 to +100 °C
Documents
Specifications
- Connection:BNC
- Ø×L:12×150 mm
- For:Aqueous media, beverage, Tris buffer
- Type:Sealed, double junction (Pellon)
- Material:Glass
- Seq:05
- Electrolyte:Gel
- pH:0 - 14
Accessories
Product Information
- Catalog #PkAvailabilityPrice
- Refill solution 3,5 mol/l KCl



