Order Entry
Export
Orders LinkContactUsLinkComponent
Interchangeable Blocks for PHMT Thermoshakers

Interchangeable Blocks for PHMT Thermoshakers

Spare block for PSC18, 20× 0,5 ml and 12×1,5 ml microtubes
Catalog # 444-0351
Supplier:   GRANT INSTRUMENTS
Interchangeable Blocks for PHMT Thermoshakers
Catalog # 444-0351
Supplier:  GRANT INSTRUMENTS
Interchangeable Blocks for PHMT Thermoshakers
Interchangeable Blocks for PHMT Thermoshakers

Spare block for PSC18, 20× 0,5 ml and 12×1,5 ml microtubes

Price
371,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Spare block for PSC18
  • Seq:
    02
  • For:
    20× 0,5 ml and 12×1,5 ml microtubes