pH buffers, DuraCal
12,00, ±0,05, Colourless, Plastic bottle, 500 mL
Supplier: HAMILTON BONADUZ
Jump to:
Product Details & Documents
For full information concerning health and safety data please contact Avantor or Hamilton directly.
- Label includes expiration date, batch number and actual value
- Traceable to NIST and PTB, certificates available to download
- Stable solutions
- Certified pH value from a DAkkS laboratory accredited for pH measurement
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Colour:Colourless
- pH accuracy:±0,05
- pH value:12,00



