Volumetric flasks, class A, white markings
50 ml, 12/21, 0,06 ± ml
WITG3.670.005.P
Borosilicate glass 3.3 amber, with PE stopper.
- White markings
- Conformity certified (DE-M marking)
- Inclusive batch certificate
: DIN EN ISO 1042
- NS:12/21
- Capacity:50 ml
- Seq:02
- Tolerance:0,06 ± ml