Orders LinkContactUsLinkComponent
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
20 ml, 10/19, 0,04 ± ml
Catalog # WITG3.670.002.P
Supplier:   witeg Labortechnik
Volumetric flasks, class A, white markings
Catalog # WITG3.670.002.P
Supplier:  witeg Labortechnik
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
20 ml, 10/19, 0,04 ± ml
Price
37,20 €
Log in for availability
1 * 2 items
Product Details & Documents
Borosilicate glass 3.3 amber, with PE stopper.
  • White markings
  • Conformity certified (DE-M marking)
  • Inclusive batch certificate
 : DIN EN ISO 1042
Specifications
  • NS:
    10/19
  • Capacity:
    20 ml
  • Seq:
    01
  • Tolerance:
    0,04 ± ml