Orders LinkContactUsLinkComponent
Sulfonation flasks, with three necks
Sulfonation flasks, with three necks
100 ml, 29/32 NS, 14/23 NS
Catalog # WITG0.851.101
Supplier:   witeg Labortechnik
Sulfonation flasks, with three necks
Catalog # WITG0.851.101
Supplier:  witeg Labortechnik
Sulfonation flasks, with three necks
Sulfonation flasks, with three necks
100 ml, 29/32 NS, 14/23 NS
Price
101,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With one parallel and two angled side necks.
Specifications
  • Capacity:
    100 ml
  • Seq:
    01
  • Side necks:
    14/23 NS
  • Centre neck:
    29/32 NS