Order Entry
Export
Orders LinkContactUsLinkComponent
Pasteur pipettes, disposable, LDPE

Pasteur pipettes, disposable, LDPE

7,3 ml, 300 mm, - ml
Catalog # SEMA4208
Supplier:   SEMADENI
Pasteur pipettes, disposable, LDPE
Catalog # SEMA4208
Supplier:  SEMADENI
Pasteur pipettes, disposable, LDPE
Pasteur pipettes, disposable, LDPE

7,3 ml, 300 mm, - ml

Price
85,00 €
Log in for availability
1 * 100 items
Product Details & Documents

LDPE, disposable, non-sterile.

  • Almost unbreakable transfer pipettes
  • Good chemical resistance and can be used at low temperatures
  • Pipettes are made from one piece: No risk of leakage
  • Dustproof packaged in convenient standard boxes
Specifications
  • Length:
    300 mm
  • Seq:
    04
  • Graduation:
    - ml
  • Bulb draw:
    7,3 ml