Order Entry
Export
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

140 mm, 34 mm, 450 mm
Catalog # SEMA0203
Supplier:   SEMADENI
Funnels with short stem
Catalog # SEMA0203
Supplier:  SEMADENI
Funnels with short stem
Funnels with short stem

140 mm, 34 mm, 450 mm

Price
69,10 €
Log in for availability
1 * 1 items
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

Funnels, made of HDPE, with smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.

  • Easy to clean
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Funnel O-Ø:
    450 mm
  • Stem O-Ø:
    34 mm
  • Stem length:
    140 mm