Orders LinkContactUsLinkComponent
Accessories for comparators
Accessories for comparators
AF332 EP colour scale, 4/64A, R1 - R7
Catalog # TINT246410
Supplier:   Lovibond Tintometer
Accessories for comparators
Catalog # TINT246410
Accessories for comparators
Accessories for comparators
AF332 EP colour scale, 4/64A, R1 - R7
Price
465,00 €
Log in for availability
1 * 1 items
Product Details & Documents
A wide range of test discs containing a series of graded glass filters, which have been pre-calibrated to give direct test readings. The unique feature of Lovibond® test discs is that they incorporate coloured glass filters which are colour stable and not affected by UV light or extreme environmental conditions.
  • Disc codes starting with 'N' are for use with the Nessleriser 2150
  • Disc codes starting with 'C' are for use with the Nessleriser 2250
  • All other discs are for use with the Comparator 2000+
Specifications
  • Range:
    R1 - R7
  • Seq:
    83
  • Code:
    4/64A
  • Test method:
    AF332 EP colour scale