Glass beads
Glass beads, solid, 7 mm
Catalog # 201-0281
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Solid glass.
For packing distillation columns.
Specifications
- Ø:7 mm
- Description:Glass beads, solid
- Seq:08
Solid glass.
For packing distillation columns.