0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Dissection Needles
Dissecting needles, with plastic handle
Dissecting needles, with plastic handle
Straight
631-7159
:
RSG
Dissecting needles, with plastic handle
631-7159
:
RSG
Dissecting needles, with plastic handle
Straight
41,10 €
1 * 10 items
-
+
Stainless steel needles with black plastic handle.
Form:
Straight