Orders LinkContactUsLinkComponent
Pharmacist's spatulas
Pharmacist's spatulas
Flexible blade, 150 mm, 18/22 mm
Catalog # RSGA051.150
Supplier:   RSG
Pharmacist's spatulas
Catalog # RSGA051.150
Supplier:  RSG
Pharmacist's spatulas
Pharmacist's spatulas
Flexible blade, 150 mm, 18/22 mm
Price
77,10 €
Log in for availability
1 * 10 items
Product Details & Documents
Pharmacist's spatulas with stainless steel blade and black plastic handle.
  • Conical blade
  • Dishwasher-proof
  • Available flexible or non flexible
Specifications
  • Description:
    Flexible blade
  • Blade width:
    18/22 mm
  • Blade length:
    150 mm