Order Entry
Export
Orders LinkContactUsLinkComponent
Double spatulas, micro, one conical end

Double spatulas, micro, one conical end

140 mm, 7 mm
Catalog # RSGA039.140
Supplier:   RSG
Double spatulas, micro, one conical end
Catalog # RSGA039.140
Supplier:  RSG
Double spatulas, micro, one conical end
Double spatulas, micro, one conical end

140 mm, 7 mm

Price
40,50 €
Log in for availability
1 * 5 items
Product Details & Documents

Micro double spatulas with one conical end, 7 mm wide. Made of stainless steel.

Specifications
  • Length:
    140 mm
  • Blade width:
    7 mm