Orders LinkContactUsLinkComponent
Bunsen burners
Bunsen burners
Bunsen burner, with air regulation and needle valve, Natural gas
Catalog # RSGA253.130
Supplier:   RSG
Bunsen burners
Catalog # RSGA253.130
Supplier:  RSG
Bunsen burners
Bunsen burners
Bunsen burner, with air regulation and needle valve, Natural gas
Price
109,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Bunsen burner made of stainless steel.
  • With air regulation sleeve
Specifications
  • Description:
    Bunsen burner, with air regulation and needle valve
  • For:
    Natural gas