Order Entry
Export
Orders LinkContactUsLinkComponent
Pharmacist's spatulas

Pharmacist's spatulas

Plastic handle, dishwasher-safe, 75 mm, 18 mm
Catalog # 231-0555
Supplier:   RSG
Pharmacist's spatulas
Catalog # 231-0555
Supplier:  RSG
Pharmacist's spatulas
Pharmacist's spatulas

Plastic handle, dishwasher-safe, 75 mm, 18 mm

Price
73,30 €
Log in for availability
1 * 10 items
Product Details & Documents

Chromium steel, hardened, extremely flexible blade.

  • Excellent thermal resistance
  • Good chemical resistance
  • Quick and easy maintenance
  • Available with wooden handle or with dishwasher-safe plastic handle
Specifications
  • Description:
    Plastic handle, dishwasher-safe
  • Blade width:
    18 mm
  • Blade length:
    75 mm