Orders LinkContactUsLinkComponent
Forceps for cover glasses
Forceps for cover glasses
Bent, premium quality, 130 mm
Catalog # 232-0195
Supplier:   RSG
Forceps for cover glasses
Catalog # 232-0195
Supplier:  RSG
Forceps for cover glasses
Forceps for cover glasses
Bent, premium quality, 130 mm
Price
63,10 €
Log in for availability
1 * 5 items
Product Details & Documents
Stainless steel.
  • For easy, secure handling of cover glasses
  • Can also be used for other laboratory items, such as filter discs
  • Easy to clean
Specifications
  • Type:
    Bent, premium quality
  • Length:
    130 mm
  • Seq:
    06