Forceps for cover glasses
Bent, premium quality, 145 mm
232-0196
: RSG
Stainless steel.
- For easy, secure handling of cover glasses
- Can also be used for other laboratory items, such as filter discs
- Easy to clean
- Type:Bent, premium quality
- Length:145 mm
- Seq:07