Pharmacist's spatulas
Hardened blade, 100 mm, 18/22 mm
Catalog # RSGA651.100
Supplier: RSG
Jump to:
Product Details & Documents
Pharmacist's spatulas with stainless steel blade and black plastic handle.
- Conical blade
- Dishwasher-proof
- Available flexible or non flexible
Specifications
- Description:Hardened blade
- Blade width:18/22 mm
- Blade length:100 mm