Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Microscopes
Accessories
Accessories
Print
Accessories
Storage and transport box
Catalog # 630-0233
Supplier:
ZEISS
Accessories
Catalog # 630-0233
Supplier:
ZEISS
Expand image
6
Return to Top
Accessories
Storage and transport box
Specifications
Add to cart
Jump to:
Specifications
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Inquire For Price
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Description:
Storage and transport box
Seq:
02