Orders LinkContactUsLinkComponent
Pipette controller, PIPETBOY acu 2
Pipette controller, PIPETBOY acu 2
PIPETBOY acu violet
Catalog # 612-2964
Supplier:   INTEGRA Biosciences
Pipette controller, PIPETBOY acu 2
Catalog # 612-2964
Pipette controller, PIPETBOY acu 2
Pipette controller, PIPETBOY acu 2
PIPETBOY acu violet
Price
473,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Pipette controller for glass and plastic pipettes from 0,1 to 100 ml.
  • Lightweight (195 g), enhanced ergonomics for stress and fatigue-free pipetting
  • Pipetting speed is conveniently pre-set via thumb wheel; speed is then easily regulated with the tips of the fingers
  • Autoclavable nose piece and easy to replace hydrophobic filter for reliable protection
  • LED provides clear information about the battery charging status
  • Colour coding for easy identification of the right controller
  • Maximum pipetting speed: 13,5 ml/s (with 50 ml pipette)
The second generation of PIPETBOY acu is equipped with the latest technology providing increased pipetting speed and significant instrument weight reduction. These new features, combined with unchanged main benefits, provide an improved ergonomic pipetting experience.
Ordering information: Supplied with a rechargeable Li-ion battery, wall holder, mains adapter and 0,45 µm sterile filter. Additional filters (0,2 and 0,45 µm) can be ordered separately.
Documents
Specifications
  • Description:
    PIPETBOY acu violet
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Spare filter, non sterile, 0,2 μm
    Spare filter, non sterile, 0,2 μm
    INTG156607
    Log in for availability
    1 * 1 items - 8,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare filter, sterile, 0,2 μm
    Spare filter, sterile, 0,2 μm
    INTG156608
    Log in for availability
    1 * 1 items - 12,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Stand for PIPETBOY (acu, pro, comfort) and VACUBOY
    Stand for PIPETBOY (acu, pro, comfort) and VACUBOY
    INTG155065
    Log in for availability
    1 * 1 items - 101,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Battery compartment lid, red
    Battery compartment lid, red
    612-0902
    Log in for availability
    1 * 1 items - 10,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Battery compartment lid, green
    Battery compartment lid, green
    612-0901
    Log in for availability
    1 * 1 items - 10,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Battery compartment lid, classic
    Battery compartment lid, classic
    612-0777
    Log in for availability
    1 * 1 items - 10,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Battery compartment lid, blue
    Battery compartment lid, blue
    612-0903
    Log in for availability
    1 * 1 items - 10,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Wall mount for PIPETBOY acu 2
    Wall mount for PIPETBOY acu 2
    612-0694
    Log in for availability
    1 * 1 items - 36,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Battery compartment lid, purple
    Battery compartment lid, purple
    INTG155164
    Log in for availability
    1 * 1 items - 10,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Battery compartment lid, transparent
    Battery compartment lid, transparent
    612-0904
    Log in for availability
    1 * 1 items - 10,60 €
Product Information
  • Catalog #
    Availability
    Price
  • O-ring
    O-ring
    612-0784
    Log in for availability
    1 * 1 items - 8,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Nose piece housing, turquoise, incl. O-ring
    Nose piece housing, turquoise, incl. O-ring
    612-0785
    Log in for availability
    1 * 1 items - 36,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Filter rubber, silicone, for holding the filter in the nose piece
    Filter rubber, silicone, for holding the filter in the nose piece
    613-4435
    Log in for availability
    1 * 1 items - 16,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Nose piece complete, turquoise, for classic and transparent models
    Nose piece complete, turquoise, for classic and transparent models
    612-0789
    Log in for availability
    1 * 1 items - 85,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Mains adapter EU, 230 VAC
    Mains adapter EU, 230 VAC
    612-0645
    Log in for availability
    1 * 1 items - 98,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Battery, 9 V NiMH, for PIPETBOY acu (instruments with serial number below ≤1350000)
    Battery, 9 V NiMH, for PIPETBOY acu (instruments with serial number below ≤1350000)
    612-0923
    Log in for availability
    1 * 1 items - 53,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Nose piece housing, grey, incl. O-ring
    Nose piece housing, grey, incl. O-ring
    612-0703
    Log in for availability
    1 * 1 items - 36,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare filter, non sterile, 0,45 µm
    Spare filter, non sterile, 0,45 µm
    613-4432
    Log in for availability
    1 * 1 items - 8,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare filter, sterile, 0,45 μm
    Spare filter, sterile, 0,45 μm
    612-0925
    Log in for availability
    1 * 1 items - 30,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Battery, lithium polymer, for PIPETBOY acu 2 (instruments with serial number from ≥1350001)
    Battery, lithium polymer, for PIPETBOY acu 2 (instruments with serial number from ≥1350001)
    INTG155066
    Log in for availability
    1 * 1 items - 98,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Nose piece complete, grey, for green, red, blue, purple models
    Nose piece complete, grey, for green, red, blue, purple models
    INTG155070
    Log in for availability
    1 * 1 items - 85,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Pipette mount, silicone, for holding the pipette in the nose piece
    Pipette mount, silicone, for holding the pipette in the nose piece
    612-0922
    Log in for availability
    1 * 1 items - 15,60 €