ContactUsLinkComponent
Thermometer, mercury, blue reflecting, DIN12784 and similar DIN, measuring range: -10:150/1C , with standard ground joint NS14.5/2 3 , upper:11mm, lower:8mm, length: 350mm, installed length:75mm, capillary lens front, colourless, richter-top finish 1 * 1 items
Catalog # AMARD262330
Supplier:  Amarell
Thermometer, mercury, blue reflecting, DIN12784 and similar DIN, measuring range: -10:150/1C , with standard ground joint NS14.5/2 3 , upper:11mm, lower:8mm, length: 350mm, installed length:75mm, capillary lens front, colourless, richter-top finish 1 * 1 items
Catalog # AMARD262330
Supplier:  Amarell